Methyl 2-[[(1S)-1-phenylethyl]amino]acetate 98%

Lieferant: Apollo Scientific

OR955654-1G
APOSOR955654-1GEA 435 EUR
APOSOR955654-1G
Methyl 2-[[(1S)-1-phenylethyl]amino]acetate 98%
Methyl 2-[[(1S)-1-phenylethyl]amino]acetate
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18903630
CAS-Nummer: 248603-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR