3-Hydroxy-3-phenyl-propionic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-hydroxy-3-phenylpropanoate

OR955657-1G
APOSOR955657-1GEA 435 EUR
APOSOR955657-1G
3-Hydroxy-3-phenyl-propionic acid methyl ester 95%
3-Hydroxy-3-phenyl-propionic acid methyl ester
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19691249
CAS-Nummer: 7497-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR