Methyl (2r)-2,3-dihydro-1H-indole-2-carboxylate 95%

Lieferant: Apollo Scientific

OR955669-1G
APOSOR955669-1GEA 502 EUR
APOSOR955669-1G
Methyl (2r)-2,3-dihydro-1H-indole-2-carboxylate 95%
Methyl (2r)-2,3-dihydro-1H-indole-2-carboxylate
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16037973
CAS-Nummer: 293737-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR