(S)-2-Amino-pent-4-enoic acid methyl ester 97%

Lieferant: Apollo Scientific

OR955678-1G
APOSOR955678-1GEA 633 EUR
APOSOR955678-1G
(S)-2-Amino-pent-4-enoic acid methyl ester 97%
(S)-2-Amino-pent-4-enoic acid methyl ester
Formel: C₆H₁₁NO₂
Molecular Weight: 129,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19203463
CAS-Nummer: 50299-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR