Methyl quinoline-8-carboxylate 95%

Lieferant: Apollo Scientific

OR955684-1G OR955684-5G
APOSOR955684-1GEA 277 EUR
APOSOR955684-1G APOSOR955684-5G
Methyl quinoline-8-carboxylate 95%
Methyl quinoline-8-carboxylate
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06203774
CAS-Nummer: 40245-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR