Methyl 2,5-dimethyloxazole-4-carboxylate 96%

Lieferant: Apollo Scientific

OR955696-1G
APOSOR955696-1GEA 632 EUR
APOSOR955696-1G
Methyl 2,5-dimethyloxazole-4-carboxylate 96%
Methyl 2,5-dimethyloxazole-4-carboxylate
Formel: C₇H₉NO₃
Molecular Weight: 155,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23159800
CAS-Nummer: 73537-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR