Methyl-6-hydroxynicotinat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate
OR9557-5G
APOSOR9557-5GEA
29
EUR
APOSOR9557-5G
Methyl-6-hydroxynicotinat 98%
Methyl-6-hydroxynicotinat
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Schmelzpunkt: 161…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04113616 CAS-Nummer: 66171-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...