Methyl 1-phenylcyclopropane-1-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1-phenylcyclopropanecarboxylate
OR955739-1G
APOSOR955739-1GEA
369
EUR
APOSOR955739-1G
Methyl 1-phenylcyclopropane-1-carboxylate 95%
Methyl 1-phenylcyclopropane-1-carboxylate
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031860 CAS-Nummer: 6121-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...