Methyl 2-methylpyrimidine-4-carboxylate 95%
Lieferant: Apollo Scientific
OR955753-1G
APOSOR955753-1GEA
289
EUR
APOSOR955753-1G
Methyl 2-methylpyrimidine-4-carboxylate 95%
Methyl 2-methylpyrimidine-4-carboxylate
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13174780 CAS-Nummer: 73955-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...