Methyl 3-(3-chlorophenyl)propanoate 98%

Lieferant: Apollo Scientific

OR955761-1G
APOSOR955761-1GEA 435 EUR
APOSOR955761-1G
Methyl 3-(3-chlorophenyl)propanoate 98%
Methyl 3-(3-chlorophenyl)propanoate
Formel: C₁₀H₁₁ClO₂
Molecular Weight: 198,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114353
CAS-Nummer: 103040-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR