Methyl 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 95%

Lieferant: Apollo Scientific

OR955779-1G
APOSOR955779-1GEA 632 EUR
APOSOR955779-1G
Methyl 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 95%
Methyl 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Formel: C₁₀H₁₂O₂S
Molecular Weight: 196,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23713207
CAS-Nummer: 92315-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR