4-Methyl-benzenepropanoic acid methyl ester 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-(p-tolyl)propanoate
OR955781-1G
APOSOR955781-1GEA
154
EUR
APOSOR955781-1G
4-Methyl-benzenepropanoic acid methyl ester 98%
4-Methyl-benzenepropanoic acid methyl ester
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16663245 CAS-Nummer: 56955-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...