Methyl oxazole-4-carboxylate 95%
Lieferant: Apollo Scientific
OR955794-1G
APOSOR955794-1GEA
632
EUR
APOSOR955794-1G
Methyl oxazole-4-carboxylate 95%
Methyl oxazole-4-carboxylate
Formel:
C₅H₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17170048 CAS-Nummer: 170487-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...