3-Amino-5-nitrophenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-nitrobenzeneboronic acid

OR9558-1G
APOSOR9558-1GEA 423 EUR
APOSOR9558-1G
3-Amino-5-nitrophenylboronsäure 97%
3-Amino-5-nitrophenylboronsäure
Formel: C₆H₇BN₂O₄
Molecular Weight: 181,94 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08436012
CAS-Nummer: 89466-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR