Methyl benzothiophene-3-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl benzo[b]thiophene-3-carboxylate

OR955809-1G
APOSOR955809-1GEA 227 EUR
APOSOR955809-1G
Methyl benzothiophene-3-carboxylate 98%
Methyl benzothiophene-3-carboxylate
Formel: C₁₀H₈O₂S
Molecular Weight: 192.234 g/mol
Siedepunkt: 165…166 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06203855
CAS-Nummer: 22913-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR