(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 96%

Lieferant: Apollo Scientific

OR955812-1G
APOSOR955812-1GEA 62.5 EUR
APOSOR955812-1G
(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 96%
(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106006
CAS-Nummer: 180323-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR