3-Methyl-4-isoxazolecarboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

OR955818-1G
APOSOR955818-1GEA 502 EUR
APOSOR955818-1G
3-Methyl-4-isoxazolecarboxylic acid methyl ester 95%
3-Methyl-4-isoxazolecarboxylic acid methyl ester
Formel: C₆H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12032284
CAS-Nummer: 92234-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR