Methyl (2R)-2-phenylpropanoate 95%
Lieferant: Apollo Scientific
Synonyme:
(R)-Methyl 2-phenylpropanoate
OR955823-1G
APOSOR955823-1GEA
260
EUR
APOSOR955823-1G
Methyl (2R)-2-phenylpropanoate 95%
Methyl (2R)-2-phenylpropanoate
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164.201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20483795 CAS-Nummer: 34083-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...