Methyl (2s)-2-amino-3-(2-methylphenyl)propanoate 95%

Lieferant: Apollo Scientific

OR955834-1G
APOSOR955834-1GEA 632 EUR
APOSOR955834-1G
Methyl (2s)-2-amino-3-(2-methylphenyl)propanoate 95%
Methyl (2s)-2-amino-3-(2-methylphenyl)propanoate
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08058210
CAS-Nummer: 1213324-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR