Methyl a-methylcinnamate 95%

Lieferant: Apollo Scientific

OR955855-1G
APOSOR955855-1GEA 632 EUR
APOSOR955855-1G
Methyl a-methylcinnamate 95%
Methyl a-methylcinnamate
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00054163
CAS-Nummer: 25692-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR