Methyl 2-[4-(([(tert-butoxy)carbonyl]amino)methyl)phenyl]acetate 98%

Lieferant: Apollo Scientific
Achtung

OR955860-250MG
APOSOR955860-250MGEA 178 EUR
APOSOR955860-250MG
Methyl 2-[4-(([(tert-butoxy)carbonyl]amino)methyl)phenyl]acetate 98%
Methyl 2-[4-(([(tert-butoxy)carbonyl]amino)methyl)phenyl]acetate
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953088
CAS-Nummer: 191871-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR