Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

OR955865-1G
APOSOR955865-1GEA 661 EUR
APOSOR955865-1G
Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride 98%
Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride
Formel: C₆H₈N₂O₂·ClH
Molecular Weight: 176,601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275082
CAS-Nummer: 51718-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR