Methyl (2s)-2-amino-3-(furan-2-yl)propanoate 95%

Lieferant: Apollo Scientific

Synonyme: MEthyl (2s)-2-amino-3-(furan-2-yl)propanoate

OR955867-1G
APOSOR955867-1GEA 343 EUR
APOSOR955867-1G
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate 95%
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate
Formel: C₈H₁₁NO₃
Molecular Weight: 169,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565764
CAS-Nummer: 146725-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR