methyl 2-(6-methoxypyridin-3-yl)acetate 96%

Lieferant: Apollo Scientific

OR955901-1G
APOSOR955901-1GEA 806 EUR
APOSOR955901-1G
methyl 2-(6-methoxypyridin-3-yl)acetate 96%
methyl 2-(6-methoxypyridin-3-yl)acetate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26520887
CAS-Nummer: 943541-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR