Methyl 2,5-dimethylthiophene-3-carboxylate 95%

Lieferant: Apollo Scientific

OR955944-1G
APOSOR955944-1GEA 806 EUR
APOSOR955944-1G
Methyl 2,5-dimethylthiophene-3-carboxylate 95%
Methyl 2,5-dimethylthiophene-3-carboxylate
Formel: C₈H₁₀O₂S
Molecular Weight: 170,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23714100
CAS-Nummer: 53562-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR