Methyl-4-chloro-1H-pyrazole-3-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2H-pyrazole-3-carboxylic acid methyl ester, Methyl 4-chloro-1H-pyrazole-3-carboxylate

OR955959-1G
APOSOR955959-1GEA 39.3 EUR
APOSOR955959-1G
Methyl-4-chloro-1H-pyrazole-3-carboxylate 95%
Methyl-4-chloro-1H-pyrazole-3-carboxylate
Formel: C₅H₅ClN₂O₂
Molecular Weight: 160,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969136
CAS-Nummer: 1005584-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR