Methyl 4-chloro-3-(chlorosulphonyl)benzoate 96%

Lieferant: Apollo Scientific
Gefahr

OR955980-1G OR955980-250MG OR955980-5G
APOSOR955980-1GEA 0 EUR
APOSOR955980-1G APOSOR955980-250MG APOSOR955980-5G
Methyl 4-chloro-3-(chlorosulphonyl)benzoate 96%
Methyl 4-chloro-3-(chlorosulphonyl)benzoate
Formel: C₈H₆Cl₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD09816501
CAS-Nummer: 1000933-19-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR