Methyl imidazo[1,2-a]pyridine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR956008-1G
APOSOR956008-1GEA 632 EUR
APOSOR956008-1G
Methyl imidazo[1,2-a]pyridine-2-carboxylate 95%
Methyl imidazo[1,2-a]pyridine-2-carboxylate
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12912213
CAS-Nummer: 1220397-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR