Methyl (2R)-2-aminopent-4-ynoate 97%
Lieferant: Apollo Scientific
OR956016-1G
OR956016-250MG
APOSOR956016-1GEA
806
EUR
APOSOR956016-1G
APOSOR956016-250MG
Methyl (2R)-2-aminopent-4-ynoate 97%
Methyl (2R)-2-aminopent-4-ynoate
Formel:
C₆H₉NO₂ Molecular Weight: 127.141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18252702 CAS-Nummer: 78342-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...