4-Methyl-1,3-benzoxazol-2-amine 95%

Lieferant: Apollo Scientific

OR956057-1G
APOSOR956057-1GEA 719 EUR
APOSOR956057-1G
4-Methyl-1,3-benzoxazol-2-amine 95%
4-Methyl-1,3-benzoxazol-2-amine
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11844835
CAS-Nummer: 947504-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR