Methyl 2-amino-1,3-benzoxazole-6-carboxylate 95%

Lieferant: Apollo Scientific

OR956060-1G
APOSOR956060-1GEA 719 EUR
APOSOR956060-1G
Methyl 2-amino-1,3-benzoxazole-6-carboxylate 95%
Methyl 2-amino-1,3-benzoxazole-6-carboxylate
Formel: C₉H₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18917418
CAS-Nummer: 851075-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR