4-(Chlorocarbonyl)-benzoic acid 95%

Lieferant: Apollo Scientific

OR956112-1G
APOSOR956112-1GEA 644 EUR
APOSOR956112-1G
4-(Chlorocarbonyl)-benzoic acid 95%
4-(Chlorocarbonyl)-benzoic acid
Formel: C₈H₅ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06208024
CAS-Nummer: 18708-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR