3-tert-Butyl-1H-pyrazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR956146-1G
APOSOR956146-1GEA 632 EUR
APOSOR956146-1G
3-tert-Butyl-1H-pyrazole-4-carboxylic acid 95%
3-tert-Butyl-1H-pyrazole-4-carboxylic acid
Formel: C₈H₁₂N₂O₂
Molecular Weight: 168,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258851
CAS-Nummer: 1152540-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR