4-(Methanesulphonylmethyl)benzonitrile 98%

Lieferant: Apollo Scientific

OR956194-1G OR956194-5G
APOSOR956194-1GEA 179 EUR
APOSOR956194-1G APOSOR956194-5G
4-(Methanesulphonylmethyl)benzonitrile 98%
4-(Methanesulphonylmethyl)benzonitrile
Formel: C₉H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD15201679
CAS-Nummer: 161417-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR