3-(Methanesulphonylmethyl)aniline 98%

Lieferant: Apollo Scientific

OR956201-1G
APOSOR956201-1GEA 168 EUR
APOSOR956201-1G
3-(Methanesulphonylmethyl)aniline 98%
3-(Methanesulphonylmethyl)aniline
Formel: C₈H₁₁NO₂S
Molecular Weight: 185.243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12776627
CAS-Nummer: 261925-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR