2-(Methanesulphonylmethyl)aniline 95%

Lieferant: Apollo Scientific

OR956238-1G
APOSOR956238-1GEA 221 EUR
APOSOR956238-1G
2-(Methanesulphonylmethyl)aniline 95%
2-(Methanesulphonylmethyl)aniline
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12823721
CAS-Nummer: 25195-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR