(2-Nitroethenyl)cyclopropane 95%

Lieferant: Apollo Scientific

OR956249-1G
APOSOR956249-1GEA 806 EUR
APOSOR956249-1G
(2-Nitroethenyl)cyclopropane 95%
(2-Nitroethenyl)cyclopropane
Formel: C₅H₇NO₂
Molecular Weight: 113,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16707055
CAS-Nummer: 1175052-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR