5-Nitro-1-propyl-1H-pyrazole 95%
Lieferant: Apollo Scientific
OR956255-1G
APOSOR956255-1GEA
538
EUR
APOSOR956255-1G
5-Nitro-1-propyl-1H-pyrazole 95%
5-Nitro-1-propyl-1H-pyrazole
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09468158 CAS-Nummer: 1170522-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...