1-(Ethylthio)-4-nitrobenzene 95%

Lieferant: Apollo Scientific

OR956269-1G
APOSOR956269-1GEA 328 EUR
APOSOR956269-1G
1-(Ethylthio)-4-nitrobenzene 95%
1-(Ethylthio)-4-nitrobenzene
Formel: C₈H₉NO₂S
Molecular Weight: 183.228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00460618
CAS-Nummer: 7205-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR