1-Cyclopentyl-5-nitro-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956274-1G
APOSOR956274-1GEA 632 EUR
APOSOR956274-1G
1-Cyclopentyl-5-nitro-1H-pyrazole 95%
1-Cyclopentyl-5-nitro-1H-pyrazole
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701860
CAS-Nummer: 1170836-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR