1-Nitro-2-vinyl-benzene 95%

Lieferant: Apollo Scientific

OR956278-1G
APOSOR956278-1GEA 952 EUR
APOSOR956278-1G
1-Nitro-2-vinyl-benzene 95%
1-Nitro-2-vinyl-benzene
Formel: C₈H₇NO₂
Molecular Weight: 149,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08461670
CAS-Nummer: 579-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR