2-Methyl-3-nitrobenzonitrile 97%

Lieferant: Apollo Scientific

OR956279-5G
APOSOR956279-5GEA 55.1 EUR
APOSOR956279-5G
2-Methyl-3-nitrobenzonitrile 97%
2-Methyl-3-nitrobenzonitrile
Formel: C₈H₆N₂O₂
Molecular Weight: 162.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025714
CAS-Nummer: 71516-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR