4-Nitroquinoline 95%

Lieferant: Apollo Scientific

OR956345-1G
APOSOR956345-1GEA 806 EUR
APOSOR956345-1G
4-Nitroquinoline 95%
4-Nitroquinoline
Formel: C₉H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01685427
CAS-Nummer: 3741-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR