N-[(2-Nitrophenyl)methyl]cyclopropanamine 95%

Lieferant: Apollo Scientific

OR956358-1G
APOSOR956358-1GEA 538 EUR
APOSOR956358-1G
N-[(2-Nitrophenyl)methyl]cyclopropanamine 95%
N-[(2-Nitrophenyl)methyl]cyclopropanamine
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09812611
CAS-Nummer: 884501-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR