Methyl (4-nitrophenyl)acetate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(4-nitrophenyl)acetate

OR956383-1G OR956383-25G OR956383-5G OR956383-250MG OR956383-10G
APOSOR956383-1GEA 48 EUR
APOSOR956383-1G APOSOR956383-25G APOSOR956383-5G APOSOR956383-250MG APOSOR956383-10G
Methyl (4-nitrophenyl)acetate 95%
Methyl (4-nitrophenyl)acetate
Formel: C₉H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00024802
CAS-Nummer: 2945-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR