(2E)-3-(4-Nitrophenyl)acrylonitrile 95%

Lieferant: Apollo Scientific

OR956391-1G
APOSOR956391-1GEA 487 EUR
APOSOR956391-1G
(2E)-3-(4-Nitrophenyl)acrylonitrile 95%
(2E)-3-(4-Nitrophenyl)acrylonitrile
Formel: C₉H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00094456
CAS-Nummer: 29246-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR