3-Nitroquinoline 95%

Lieferant: Apollo Scientific

OR956394-1G
APOSOR956394-1GEA 647 EUR
APOSOR956394-1G
3-Nitroquinoline 95%
3-Nitroquinoline
Formel: C₉H₆N₂O₂
Molecular Weight: 174,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00956379
CAS-Nummer: 17576-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR