5-Bromo-1-ethyl-1H-pyrazole 95%
Lieferant: Apollo Scientific
OR956652-1G
APOSOR956652-1GEA
647
EUR
APOSOR956652-1G
5-Bromo-1-ethyl-1H-pyrazole 95%
5-Bromo-1-ethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18447684 CAS-Nummer: 1268334-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...