3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR956691-1G
APOSOR956691-1GEA 632 EUR
APOSOR956691-1G
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile 95%
3-Bromo-1,5-dimethyl-1H-pyrrole-2-carbonitrile
Formel: C₇H₇BrN₂
Molecular Weight: 199,048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21336851
CAS-Nummer: 942612-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR