2-Chloro-furan 95%

Lieferant: Apollo Scientific

OR956904-1G
APOSOR956904-1GEA 632 EUR
APOSOR956904-1G
2-Chloro-furan 95%
2-Chloro-furan
Formel: C₄H₃ClO
Molecular Weight: 102.519 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143721
CAS-Nummer: 3187-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR